ContactUsLinkComponent
EasySep™ Human Resting CD4+ T Cell Isolation Kits
EasySep™ Human Resting CD4+ T Cell Isolation Kits
  MSPP-17962
undefined
EasySep™ Human Resting CD4+ T Cell Isolation Kits
  MSPP-17962
 :  17962
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ human resting CD4+ T cell isolation kit
  • Cat. No.:
    MSPP-17962
Immunomagnetic isolation of untouched human resting CD4+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').