ContactUsLinkComponent
EasySep™ Human Naïve CD8+ T Cell Isolation Kit II
EasySep™ Human Naïve CD8+ T Cell Isolation Kit II
Catalog # MSPP-17968
undefined
EasySep™ Human Naïve CD8+ T Cell Isolation Kit II
Catalog # MSPP-17968
Supplier Number:  17968
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ human naive CD8+ T cell isolation kit II
  • Cat. No.:
    MSPP-17968
Specifications
About this item
Immunomagnetic isolation of untouched human naïve CD8+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit II is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically-labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product replaces the EasySep™ Human Naïve CD8+ T Cell Enrichment Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').