ContactUsLinkComponent
EasySep™ Human Plasmacytoid DC Isolation Kits
EasySep™ Human Plasmacytoid DC Isolation Kits
  MSPP-17977
undefined
EasySep™ Human Plasmacytoid DC Isolation Kits
  MSPP-17977
 :  17977
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ human plasmacytoid DC isolation kit
  • Cat. No.:
    MSPP-17977
Immunomagnetic negative selection kit.
  • Fast, easy-to-use, and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit, for even faster cell isolations and is compatible with a greater number of EasySep™ magnets.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').