ContactUsLinkComponent
EasySep™ Mouse CD117 (cKits) Positive Selection Kits
EasySep™ Mouse CD117 (cKits) Positive Selection Kits
Catalog # MSPP-18757
undefined
EasySep™ Mouse CD117 (cKits) Positive Selection Kits
Catalog # MSPP-18757
Supplier Number:  18757
Specifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ mouse CD117 (cKIT) positive selection kit
  • Cat. No.:
    MSPP-18757
Specifications
About this item
Immunomagnetic positive selection of CD117+ cells (e.g. hematopoietic stem/progenitor cells) from mouse bone marrow.
  • Fast and easy-to-use
  • Up to 95% purity
  • No columns required
The EasySep™ Mouse CD117 (cKIT) Positive Selection Kit is designed to isolate CD117+ cells from single-cell suspensions of bone marrow by positive selection. The cocktail contains a combination of PE-labeled antibodies that is directed against CD117, which are then recognized by antibody complexes that are directed against PE and dextran. Labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').