ContactUsLinkComponent
EasySep™ Mouse CD4 Positive Selection Kit II
EasySep™ Mouse CD4 Positive Selection Kit II
Catalog # MSPP-18952
undefined
EasySep™ Mouse CD4 Positive Selection Kit II
Catalog # MSPP-18952
Supplier Number:  18952
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ mouse CD4 positive selection kit II
  • Cat. No.:
    MSPP-18952
Specifications
About this item
Immunomagnetic positive selection of CD4+ T cells from mouse splenocytes or other tissues.
  • Fast, easy-to-use, and column-free
  • Up to 98% purity
  • Isolated cells are not fluorochrome-labeled
The EasySep™ Mouse CD4 Positive Selection Kit II is designed to isolate CD4+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognizing CD4 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Mouse CD4 Positive Selection Kit for even faster cell isolations and does not result in the labeling of isolated cells with PE.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').