ContactUsLinkComponent
EasySep™ Mouse CD8a Positive Selection Kit II
EasySep™ Mouse CD8a Positive Selection Kit II
Catalog # MSPP-18953
undefined
EasySep™ Mouse CD8a Positive Selection Kit II
Catalog # MSPP-18953
Supplier Number:  18953
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ mouse CD8a positive selection kit II
  • Cat. No.:
    MSPP-18953
Specifications
About this item
Immunomagnetic positive selection of CD8a+ cells from mouse splenocytes or other tissues.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Isolated cells are not fluorochrome labeled
The EasySep™ Mouse CD8a Positive Selection Kit II is designed to isolate CD8a+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognizing CD8a and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Mouse CD8a Positive Selection Kit for even faster cell isolations and does not result in the labeling of isolated cells with PE.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').