Specifications
- Capacity:2×10⁹ cells
- Description:EasySep™ mouse CD19 positive selection kit II
- Cat. No.:MSPP-18954
- Supplier no.:18954
Specifications
About this item
Immunomagnetic positive selection of CD19+ cells (e.g. B cells) from mouse splenocytes or other tissues.
- Fast and easy-to-use
- Up to 99% purity
- No columns required
- Isolated cells are not fluorochrome-labeled
The EasySep™ Mouse CD19 Positive Selection Kit II is designed to isolate CD19+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognizing CD19 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Mouse CD19 Positive Selection Kit for even faster cell isolations and does not result in the labeling of isolated cells with PE.