Specifications
- Capacity:2×10⁹ cells
- Description:EasySep™ mouse CD90.1 positive selection kit
- Cat. No.:MSPP-18958
Specifications
About this item
Immunomagnetic positive selection of CD90.1+ cells from mouse splenocytes, lymph nodes or from peripheral blood.
- Fast, and easy-to-use
- Up to 97% purity
- No columns required
- Isolated cells are not fluorochrome-labeled
The EasySep™ Mouse CD90.1 Positive Selection Kit is designed to isolate CD90.1+ (Thy1.1+) cells from single suspensions of splenocytes, lymph nodes and peripheral blood by positive selection. Desired cells are targeted with antibody complexes recognizing CD90.1 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.