To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:60 ml buffy coat or whole blood processed
- Description:EasySep™ HLA chimerism whole blood CD19 positive selection kit
- Cat. No.:MSPP-17874
Specifications
About this item
Column-free immunomagnetic positive selection of human CD19+ cells for HLA analysis
- Fast and easy-to-use
- Up to 99% purity
- No columns required
- Compatible across EasySep™ 'The Big Easy', EasyEights™, and RoboSep™-S platforms
The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.
This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations.