ContactUsLinkComponent
EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kits
EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kits
Catalog # MSPP-19232
undefined
EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kits
Catalog # MSPP-19232
Supplier Number:  19232
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ human CD4+CD127lowCD49d- regulatory T cell enrichment kit
  • Cat. No.:
    MSPP-19232
Specifications
About this item
Immunomagnetic isolation of untouched human CD4+CD127lowCD49d- T cells
  • Fast, easy-to-use and column-free
  • Up to 87.4% purity
  • Isolated cells are untouched
The EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kit is designed to isolate CD4+CD127lowCD49d- cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells, CD127high, CD49d+ cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').