Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ non-human primate T cell isolation kit
- Cat. No.:MSPP-19581
Specifications
About this item
Immunomagnetic isolation of untouched non-human primate T cells
- Fast, easy to use, and column-free
- Up to 90% purity with high recovery
- Untouched, viable cells
The EasySep™ Non-Human Primate T Cell Isolation Kit is designed to isolate T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit has been verified for use with rhesus and cynomolgus macaques.