EasySep™ Non-Human Primate CD4+ T Cell Isolation Kits
MSPP-19582
- Capacity:1×10⁹ cells
- Description:EasySep™ non-human primate CD4+ T cell isolation kit
- Cat. No.:MSPP-19582
Immunomagnetic isolation of untouched non-human primate CD4+ T cells
- Fast, easy to use, and column-free
- Up to 85% purity with high recovery
- Untouched, viable cells
The EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit has been verified for use with rhesus and cynomolgus macaques.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').