About this item
Immunomagnetic isolation of untouched human peripheral blood mononuclear cells from whole blood
- 99.9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
- Fast, easy-to-use and column-free
- Isolated cells are untouched
The EasySep™ Direct Human PBMC Isolation Kit is designed to isolate highly purified peripheral blood mononuclear cells (PBMCs) from fresh whole blood, buffy coat, bone marrow, cord blood, or leukapheresis products by negative selection. Red blood cells (RBCs), platelets, and unwanted cells are targeted for removal with antibody complexes and magnetic particles and separated using an EasySep™ magnet. Untouched PBMCs are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction.