EasySep™ Direct Human T Cell Isolation Kits
Catalog # MSPP-19661
Supplier: STEMCELL Technologies
Specifications
- Capacity:100 ml whole blood processed
- Description:EasySep™ direct human T cell isolation kit
- Cat. No.:MSPP-19661
Specifications
About this item
Immunomagnetic isolation of T cells directly from whole blood
- >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
- Up to 97% purity of isolated cells
- Fast, easy-to-use and column-free
- Isolated cells are untouched
This cell separation kit can be used to isolate highly purified T cells directly from human whole blood by immunomagnetic negative selection.
This kit targets non-T cells for removal with antibodies recognizing specific surface markers. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and can directly be used for functional assays, flow cytometry or DNA/RNA extraction.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').