ContactUsLinkComponent
EasySep™ Direct Human CD4+ T Cell Isolation Kits
EasySep™ Direct Human CD4+ T Cell Isolation Kits
Catalog # MSPP-19662
undefined
EasySep™ Direct Human CD4+ T Cell Isolation Kits
Catalog # MSPP-19662
Supplier Number:  19662
Specifications
  • Capacity:
    100 ml whole blood processed
  • Description:
    EasySep™ direct human CD4+ T cell isolation kit
  • Cat. No.:
    MSPP-19662
Specifications
About this item
Immunomagnetic isolation of CD4+ T cells directly from whole blood
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Up to 96% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
Isolate CD4+ T cells directly from human whole blood by immunomagnetic negative selection. Desired cells undergo minimal manipulation and are untouched and highly purified.
This product labels RBCs, platelets and unwanted cells with antibody complexes and magnetic particles. The magnetically labeled RBCs, platelets and non-target cells are separated from the untouched CD4+ T cells in an EasySep™ magnet and poured or pipetted into a new tube.
​Isolated cells are available for use in downstream uses such as flow cytometry, functional assays or culture.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').