Orders LinkContactUsLinkComponent
EasySep™ Direct Human B-CLL Cell Isolation Kit
EasySep™ Direct Human B-CLL Cell Isolation Kit
EasySep™ direct human B-CLL cell isolation kit, 100 ml whole blood processed
Catalog # MSPP-19664
Marketsource
Supplier #:  19664
EasySep™ Direct Human B-CLL Cell Isolation Kit
Catalog # MSPP-19664
Marketsource
EasySep™ Direct Human B-CLL Cell Isolation Kit
EasySep™ Direct Human B-CLL Cell Isolation Kit
EasySep™ direct human B-CLL cell isolation kit, 100 ml whole blood processed
Price
$1,075.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Immunomagnetic isolation of B-CLL cells directly from whole blood
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
This kit isolates untouched and highly purified B-CLL cells directly from human whole blood by fully immunomagnetic negative selection. The depletion cocktail in this kit does not contain antibodies against CD43, which can be expressed on malignant cells in chronic lymphocytic leukemia (CLL) samples.
This cell isolation kit targets non-B cells for removal based on specific surface antigens. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and can immediately be used in downstream applications such as nucleic acid extraction, flow cytometry or culture.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
  • Capacity:
    100 ml whole blood processed
  • Description:
    EasySep™ direct human B-CLL cell isolation kit