- Capacity:100 ml whole blood processed
- Description:EasySep™ direct human monocyte isolation kit
- Cat. No.:MSPP-19669
Immunomagnetic isolation of monocytes directly from whole blood
- 99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
- Up to 92% purity of isolated cells
- Fast, easy-to-use and column-free
- Isolated cells are untouched
The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately available for flow cytometry, functional assays, culture and other downstream applications.