ContactUsLinkComponent
EasySep™ Direct Human B Cell Isolation Kits
EasySep™ Direct Human B Cell Isolation Kits
Catalog # MSPP-19674
undefined
EasySep™ Direct Human B Cell Isolation Kits
Catalog # MSPP-19674
Supplier Number:  19674
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    100 ml whole blood processed
  • Description:
    EasySep™ direct human B cell isolation kit
  • Cat. No.:
    MSPP-19674
Specifications
About this item
Immunomagnetic isolation of B cells directly from whole blood
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 95% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
Isolate highly purified B cells directly from human whole blood without the need for density gradient centrifugation, RBC sedimentation or lysis. This platform provides fast and fully immunomagnetic negative selection.
This cell isolation kit targets non-B cells for removal with EasySep™ Direct RapidSpheres™ and antibodies recognizing specific surface antigens. Unwanted cells are separated from desired cells using an EasySep™ magnet, and untouched target cells are then pipetted or poured into a new tube.
Isolated cells can be used directly in downstream applications such as flow cytometry, culture or DNA/RNA extraction.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').