Specifications
- Capacity:2×10⁹ cells
- Description:EasySep™ mouse plasmacytoid DC isolation kit
- Cat. No.:MSPP-19764
Specifications
About this item
Immunomagnetic isolation of untouched mouse plasmacytoid dendritic cells (pDC)
- Fast, easy-to-use and column-free
- Up to 94% purity
- Isolated cells are untouched
The EasySep™ Mouse Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-pDCs. Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran. These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.