- Capacity:1×10⁹ cells
- Description:EasySep™ mouse memory CD4+ T cell isolation kit
- Cat. No.:MSPP-19767
- Supplier no.:19767
Immunomagnetic isolation of untouched mouse memory CD4 T cells
- Fast, easy-to-use and column-free
- Up to 96% purity
- Untouched, viable cells
The EasySep™ Mouse Memory CD4+ T Cell Isolation Kit is designed to isolate pan-memory CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-memory CD4+ T cells , featuring RapidSphere™ technology.