Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ mouse pan-B cell isolation kit
- Cat. No.:MSPP-19844
- Supplier no.:19844
Specifications
About this item
Immunomagnetic isolation of untouched mouse pan-B cells
- Fast and easy-to-use
- Up to 98% purity
- No columns required
- Untouched, viable cells
The EasySep™ Mouse Pan-B Cell Isolation Kit is designed to isolate all B cells, including conventional B-2 B cells, B-1 B cells and plasma cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non pan-B cells to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.