ContactUsLinkComponent
EasySep™ Mouse Pan-Naïve T Cell Isolation Kits
EasySep™ Mouse Pan-Naïve T Cell Isolation Kits
  MSPP-19848
undefined
EasySep™ Mouse Pan-Naïve T Cell Isolation Kits
  MSPP-19848
 :  19848
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ mouse pan-Naïve T cell isolation kit
  • Cat. No.:
    MSPP-19848
Immunomagnetic isolation of untouched mouse pan-naïve T cells
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Pan-Naïve T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve T cells to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').