Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ mouse B cell isolation kit
- Cat. No.:MSPP-19854
- Supplier no.:19854
Specifications
About this item
Immunomagnetic isolation of untouched mouse B cells
- Fast and easy-to-use
- Up to 95% purity
- No columns required
- Untouched, viable cells
The EasySep™ Mouse B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-B cells and streptavidin-coated magnetic particles).
This product replaces the EasySep™ Mouse B Cell Enrichment Kit (Catalog #19754) for even faster cell isolations.