ContactUsLinkComponent
EasySep™ Mouse Monocyte Isolation Kits
EasySep™ Mouse Monocyte Isolation Kits
Catalog # MSPP-19861
undefined
EasySep™ Mouse Monocyte Isolation Kits
Catalog # MSPP-19861
Supplier Number:  19861
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ mouse monocyte isolation kit
  • Cat. No.:
    MSPP-19861
  • Supplier no.:
    19861
Specifications
About this item
Immunomagnetic isolation of untouched mouse monocytes
  • Fast and easy-to-use
  • Up to 95% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Monocyte Isolation Kit targets non-monocyte cells by labeling unwanted cells with antibodies and magnetic particles, and separates cells without columns using an EasySep™ magnet. Desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays.
This product replaces the EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').