Specifications
- Capacity:2×10⁹ cells
- Description:EasySep™ mouse Pan-DC enrichment kit II
- Cat. No.:MSPP-19863
Specifications
About this item
Immunomagnetic negative selection kit
- Fast, easy-to-use, and column-free
- Up to 65% purity
- Isolated cells are untouched
- Optimized for high yield
- Facilitates rapid flow sorting of DC subsets
The EasySep™ Mouse Pan-DC Enrichment Kit II is designed to isolate dendritic cells (DCs) from mouse splenocytes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product replaces the EasySep™ Mouse Pan-DC Enrichment Kit (Catalog #19763), for improved yield and even faster cell isolations.