EasySep™ Release Human CD45 Positive Selection Kits
About this item
Immunomagnetic positive selection of particle-free CD45+ leukocytes from primary human tissues and tumors.
- Highly purified CD45+ cells isolated in less than 45 minutes
- No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Human CD45 Positive Selection Kit is designed to isolate CD45+ leukocytes from single-cell suspensions of primary human tissues and tumors, or tissues and tumor xenografts from humanized mice, by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD45+ cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Immunomagnetic positive selection of particle-free CD45+ leukocytes from primary human tissues and tumors.
- Highly purified CD45+ cells isolated in less than 45 minutes
- No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Human CD45 Positive Selection Kit is designed to isolate CD45+ leukocytes from single-cell suspensions of primary human tissues and tumors, or tissues and tumor xenografts from humanized mice, by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD45+ cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Product Family Options
Product Information
- CapacityAvailabilityPrice
- MSPP-100-0105EasySep™ Release human CD45 positive selection kitEasySep™ Release human CD45 positive selection kit1×10⁹ cells processedLog in for availabilityEach$1,240.00Supplier Number: 100-0105Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- MSPP-100-0107Immunomagnetic positive selection of particle-free CD45+ leukocytes from tissues and tumor xenografts from humanized miceImmunomagnetic positive selection of particle-free CD45+ leukocytes from tissues and tumor xenografts from humanized mice1×10⁹ cells processedLog in for availabilityEach$1,532.31Supplier Number: 100-0107Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource