ContactUsLinkComponent
EasySep™ Release Mouse CD138 Positive Selection Kits
EasySep™ Release Mouse CD138 Positive Selection Kits
Catalog # MSPP-100-0601
undefined
EasySep™ Release Mouse CD138 Positive Selection Kits
Catalog # MSPP-100-0601
Supplier Number:  100-0601
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ Release Mouse CD138 positive selection kit
  • Cat. No.:
    MSPP-100-0601
Specifications
About this item
Immunomagnetic positive selection kit using particle release technology.
  • Up to 95% purity of plasma cells from immunized mouse samples
  • No-wash removal of EasySep™ Releasable RapidSpheres™
  • Compatible with hybridoma generation protocols, including electrofusion
Isolate magnetic particle-free CD138+ plasma cells and plasmablasts from single-cell suspensions of mouse splenocytes or bone marrow in as little as 37 minutes. CD138+ cells are differentiated B cells that can be used in antibody discovery research due to their ability to generate affinity-matured antibodies. While hybridoma fusions with mouse bone marrow and splenocytes typically require screening thousands of colonies, enriching for CD138+ plasma cells prior to hybridoma fusion can substantially decrease the screening burden to generate hybridomas.
To achieve higher lab efficiency for your antibody discovery research, purify CD138+ plasma cells and plasmablasts without columns by using immunomagnetic positive selection. Simply label cells with antibodies and magnetic particles, place in an EasySep™ cell separation magnet, then remove the bound magnetic particles from the isolated CD138+ cell fraction. CD138+ cells isolated from immunized mice are up to 95% pure and can be used with hybridoma generation protocols such as electrofusion.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).