ContactUsLinkComponent
RosetteSep™ CTC Enrichment Cocktails Containing Anti-CD36
RosetteSep™ CTC Enrichment Cocktails Containing Anti-CD36
Catalog # MSPP-15167
undefined
RosetteSep™ CTC Enrichment Cocktails Containing Anti-CD36
Catalog # MSPP-15167
Supplier Number:  15167
Specifications
  • Capacity:
    200 ml whole blood processed
  • Description:
    RosetteSep™ CTC enrichment cocktail containing anti-CD36
  • Cat. No.:
    MSPP-15167
Specifications
About this item
Immunodensity isolation of untouched circulating tumor cells.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD36 is designed to enrich circulating epithelial tumor cells from fresh whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD16, CD19, CD36, CD38, CD45, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a density gradient medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified tumor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium.
This cocktail is compatible with small-cell carcinoma samples, including lung cancer samples. If you are working with breast cancer samples, use the RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD56 (STMC15137).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).