EasySep™ Human PE Positive Selection Kit II
MSPP-17664
- Capacity:1×10⁹ cells
- Description:EasySep™ human PE positive selection kit II
- Cat. No.:MSPP-17664
Immunomagnetic positive selection of human cells labeled with PE-conjugated antibodies.
- Fast and easy-to-use
- No columns required
The EasySep™ Human PE Positive Selection Kit II is designed to isolate cells that are labeled with PE (phycoerythrin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Human PE Positive Selection Kit (STMC18551).
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).