- Capacity:1×10⁹ cells
- Description:EasySep™ APC positive selection kit II
- Cat. No.:MSPP-17681
Immunomagnetic positive selection of any cells labeled with APC-conjugated antibodies.
- Fast and easy-to-use
- No columns required
The EasySep™ APC Positive Selection Kit II is designed to isolate cells that are labeled with APC (allophycocyanin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognizing APC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ APC Positive Selection Kit (STMC18453) for even faster cell isolations.