ContactUsLinkComponent
EasySep™ Biotin Positive Selection Kit II
EasySep™ Biotin Positive Selection Kit II
Catalog # MSPP-17683
undefined
EasySep™ Biotin Positive Selection Kit II
Catalog # MSPP-17683
Supplier Number:  17683
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ Biotin positive selection kit II
  • Cat. No.:
    MSPP-17683
Specifications
About this item
Immunomagnetic positive selection of any cells labeled with biotinylated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Biotin Positive Selection Kit II is designed to isolate cells that are labeled with biotinylated antibodies. Desired cells are targeted with antibody complexes recognizing biotin and dextran-coated magnetic particles. The labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Biotin Positive Selection Kit (STMC17683) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).