ContactUsLinkComponent
EasySep™ Human EpCAM Positive Selection Kit II
EasySep™ Human EpCAM Positive Selection Kit II
Catalog # MSPP-17846
undefined
EasySep™ Human EpCAM Positive Selection Kit II
Catalog # MSPP-17846
Supplier Number:  17846
Specifications
  • Cat. No.:
    MSPP-17846
Specifications
About this item
Immunomagnetic positive selection of human EpCAM+ cells (e.g. mammary epithelial cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human EpCAM Positive Selection Kit II is designed to isolate EpCAM+ cells from fresh or previously frozen human mammary epithelial cell preparations by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognizing EpCAM and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The EpCAM+ fraction contains luminal cells, luminal-restricted, and bipotential progenitor cells.
This product replaces the EasySep™ Human EpCAM Positive Selection Kit (STMC18356).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).