Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ human CD3 positive selection kit II
- Cat. No.:MSPP-17851
Specifications
About this item
Immunomagnetic positive selection of human CD3+ cells (e.g. T cells).
- Fast and easy-to-use
- Up to 99% purity
- No columns required
The EasySep™ Human CD3 Positive Selection Kit II is designed to isolate CD3+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD3 antigen is expressed on all T cells and CD56+ NKT cells.
This product replaces the EasySep™ Human CD3 Positive Selection Kit (STMC18051) for even faster cell isolations.