ContactUsLinkComponent
EasySep™ Human CD19 Positive Selection Kit II
EasySep™ Human CD19 Positive Selection Kit II
Catalog # MSPP-17854
undefined
EasySep™ Human CD19 Positive Selection Kit II
Catalog # MSPP-17854
Supplier Number:  17854
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ human CD19 positive selection kit II
  • Cat. No.:
    MSPP-17854
Specifications
About this item
Immunomagnetic positive selection of human CD19+ cells (e.g. B cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human CD19 Positive Selection Kit II is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.
This product replaces the EasySep™ Human CD19 Positive Selection Kit (STMC18054) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).