Specifications
- Capacity:2×10⁹ cells
- Description:EasySep™ Mouse CD45 positive selection kit
- Cat. No.:MSPP-18945
Specifications
About this item
Immunomagnetic positive selection of CD45+ leukocytes from mouse lung, splenocytes, or other tissues.
- Fast and easy-to-use
- Up to 98% purity
- No columns required
- Isolated cells are not fluorochrome-labeled
The EasySep™ Mouse CD45 Positive Selection Kit is designed to isolate CD45 cells from single-cell suspensions of splenocytes, lung, or other tissues by positive selection. Desired cells are targeted with antibody complexes recognizing CD45 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.