Order Entry
Puerto Rico
ContactUsLinkComponent
Sheep IL6 ELISA Kit
Sheep IL6 ELISA Kit
Catalog # MSPP-SEA079OV
CAS Number:  
undefined
Sheep IL6 ELISA Kit
Catalog # MSPP-SEA079OV
Supplier Number:  SEA079OV
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Sheep
  • Target protein:
    IL6
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 6 (IL6) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    7.8 - 500 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    3.2 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA079OV
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Sheep IL6 (Interleukin 6). The assay range is from 7.8 to 500 pg/ml (Sandwich kit) with a sensitivity of 3.2 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

IL-6 is a multifunctional protein produced by lymphoid and non-lymphoid cells, and by normal and transformed cells, including T cells, monocyte/macrophages, fibroblasts, hepatocytes, vascular endothelial cells, cardiac myxomas, bladder cell carcinomas, myelomas, astrogliomas and glioblastomas. The production of IL-6 in these various cells is regulated, either positively or negatively, by a variety of signals including mitogens, antigenic stimulation, lipopolysaccharides, IL-1, TNF, PDGF and viruses. On the basis of its various activities, IL-6 has also been called interferon-β2 (IFN-β2), 26 kDa protein, B-cell stimulatory factor-2 (BSF-2), hybridoma/plasmacytoma growth factor, hepatocyte stimulating factor, cytotoxic T-cell differentiation factor, and macrophage-granulocyte inducing factor 2A (MGI-2A).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.