Order Entry
Puerto Rico
ContactUsLinkComponent
Rat SFTPA1 ELISA Kit
Rat SFTPA1 ELISA Kit
Catalog # MSPP-SEA890RA
CAS Number:  
undefined
Rat SFTPA1 ELISA Kit
Catalog # MSPP-SEA890RA
Supplier Number:  SEA890RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    SFTPA1
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, lung lavage fluid and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Pulmonary Surfactant Associated Protein A1 (SFTPA1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA890RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat SFTPA1 (Pulmonary Surfactant Associated Protein A1). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.5 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

SP-A (PRL) is a peptide hormone primarily associated with lactation. In breastfeeding, the infant suckling the teat stimulates the production of SP-A, which fills the breast with milk (lactogenesis) in preparation for the next feed. Oxytocin, a similar hormone is also released, which triggers milk let-down. SP-A (PRL) is a polypeptide hormone secreted by anterior pituitary of both male and female. Release of SP-A is controlled by a complex neuroendocrine reflex initiated by a tactile stimulus and regulated by hypothalamic releasing and inhibition. Genetic variations in SFTPA1 are associated with respiratory distress syndrome in premature infants (RDS) also known as RDS in prematurity. RDS in the newborn is the main cause of mortality and morbidity in premature infants.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.