Order Entry
Puerto Rico
ContactUsLinkComponent
Rat NLRP3 ELISA Kit
Rat NLRP3 ELISA Kit
Catalog # MSPP-SEK115RA
CAS Number:  
undefined
Rat NLRP3 ELISA Kit
Catalog # MSPP-SEK115RA
Supplier Number:  SEK115RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    NLRP3
  • Size:
    1 Kit
  • Sample Type:
    Tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.115 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEK115RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat NLRP3 (NLR Family Pyrin Domain Containing Protein 3). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.115 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

NLRP3 encodes a pyrin-like protein which contains a pyrin domain, a nucleotide-binding site (NBS) domain, and a leucine-rich repeat (LRR) motif. This protein interacts with apoptosis-associated speck-like protein containing a CARD. Proteins which contain the caspase recruitment domain, CARD, have been shown to be involved in inflammation and immune response. This protein may function as an activator of NF-κB signaling. The protein may play a role in the regulation of inflammation and apoptosis. Mutations in this gene have been associated with a spectrum of dominantly inherited autoinflammatory diseases called cryopyrin-associated periodic syndrome (CAPS). Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.