Order Entry
Puerto Rico
ContactUsLinkComponent
 
Rat MMP9 ELISA Kit
Catalog # MSPP-SEA553RA
CAS Number:  
undefined
Rat MMP9 ELISA Kit
Catalog # MSPP-SEA553RA
Supplier Number:  SEA553RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    MMP9
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Matrix Metalloproteinase 9 (MMP9) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    4.7 - 300 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    1.8 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA553RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat MMP9 (Matrix Metalloproteinase 9). The assay range is from 4.7 to 300 ng/ml (Sandwich kit) with a sensitivity of 1.8 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

MMP-9 (also known as gelatinase B) is secreted as a 92 kDa zymogen. Cleavage of pro-MMP-9 at or near residue 87 results in the active enzyme with a mass of approximately 82 kDa. MMP-9 has three fibronectin type II domains, a hemopexin-like domain and a proline-rich type V collagen-homologous domain. Pro-MMP-9 can be activated by MMP-3 or by certain bacterial proteinases. MMP-9 is inhibited byα2-macroglobulin or by TIMP-1, which binds to pro-MMP-9 as well as to active MMP-9.Pro-MMP-9 is secreted by monocytes, macrophages, neutrophils, keratinocytes, fibroblasts, osteoclasts, chondrocytes, skeletal muscle satellite cells, endothelial cells, and various tumor cells. Pro-MMP-9 expression is upregulated by TGF-β1, IL-1β, TGF-α, PDGF-AB,

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.