Order Entry
Puerto Rico
ContactUsLinkComponent
Rat MMP7 ELISA Kit
Rat MMP7 ELISA Kit
Catalog # MSPP-SEA102RA
CAS Number:  
undefined
Rat MMP7 ELISA Kit
Catalog # MSPP-SEA102RA
Supplier Number:  SEA102RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    MMP7
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Matrix Metalloproteinase 7 (MMP7) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.119 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA102RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat MMP7 (Matrix Metalloproteinase 7). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.119 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

MMP7, also known as Matrilysin, is the smallest member of the MMP-family, it is a zinc-dependent metalloproteinase and is overexpressed in colon cancer and many other human cancers. MMP7 is a secreted matrix metalloproteinase, which contributes to angiogenesis by breaking down basement membranes. Matrix Metalloproteinases (MMPs) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP7 is responsible for hydrolyzing proteoglycans and ECM glycoproteins. It is expressed in normal cells, specifically glandular epithelial cells. MMP7, is thought to play an important role in the early steps of tumor progression. Expression of MMP2, MMP7 and MMP11 is greater in pancreatic carcinoma than in normal pancreas.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.