Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Kim1 ELISA Kit
Rat Kim1 ELISA Kit
Catalog # MSPP-SEA785RA
Supplier:  Cloud-Clone
CAS Number:  
undefined
Rat Kim1 ELISA Kit
Catalog # MSPP-SEA785RA
Supplier:  Cloud-Clone
Supplier Number:  SEA785RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Kim1
  • Size:
    1 Kit
  • Sample Type:
    Urine, tissue homogenates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Kidney Injury Molecule 1 (Kim1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    62.5 - 4000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    24.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA785RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat Kim1 (Kidney Injury Molecule 1). The assay range is from 62.5 to 4000 pg/ml (Sandwich kit) with a sensitivity of 24.5 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

kidney injury molecule-1 (KIM-1) is a type 1 transmembrane protein that is not detectable in normal kidney tissue but is expressed at high levels in rodent kidneys with dedifferentiated proximal tubule epithelial cells after ischemic or toxic injury. Sequence analysis predicted that the 359-amino acid type I HAVCR1 glycoprotein, which is 79% identical to the monkey protein, contains a signal sequence, a 109-amino acid cys-rich N-terminal region with only 1 conserved N-glycosylation site, a 163-amino acid TSP-rich region with 3 N-glycosylation sites, 2 of which are conserved, and a 48-amino acid intracellular domain, 12 residues shorter than that of the monkey protein. Northern blot analysis revealed ubiquitous expression of a 4.4-kb HAVCR1 transcript, with highest levels in kidney and testis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.