- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Competitive
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Rat
- Target protein:FGF2
- Size:1 Kit
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Fibroblast Growth Factor 2, Basic (FGF2) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:2 h
- Shelf Life:12 Months
- Detection Range:12.35 - 1000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:50 µl
- Sensitivity:5.11 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-CEA551RA
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Rat FGF2 (Fibroblast Growth Factor 2 Basic). The assay range is from 12.35 to 1000 pg/ml (Competitive kit) with a sensitivity of 5.11 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
FGF basic is one of at least 22 mitogenic proteins of the FGF family, which show 35 to 60% amino acid conservation.Unlike other FGFs, FGF acidic and basic lack signal peptides and are secreted by an alternate pathway. Storage pools within the cell or on cell surface heparan sulfate proteoglycans (HSPG) are likely. The predicted 18 kDa FGF basic isoform can be located in both the cytoplasm and the nucleus and is presumed to be the form secreted. Transcription from alternate start sites produces 21 to 23 kDa forms found only in the nucleus. High and low molecular weight human FGF basic targets the expression of different genes when expressed in NIH-3T3 cells.The 18 kDa rat sequence has 98% aa identity with mouse, and 96 to 97% identity with human, bovine and sheep FGF basic.