Order Entry
Puerto Rico
ContactUsLinkComponent
Rat COL2 ELISA Kit
Rat COL2 ELISA Kit
Catalog # MSPP-SEA572RA
CAS Number:  
undefined
Rat COL2 ELISA Kit
Catalog # MSPP-SEA572RA
Supplier Number:  SEA572RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    COL2
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Collagen Type II (COL2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    3.12 - 200 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    1.29 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA572RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat COL2 (Collagen Type II). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.29 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Type-II collagen is the basis for articular cartilage and hyaline cartilage.It makes up 50% of all protein in cartilage and 85 to 90% of collagen of articular cartilage. Type II collagen does form fibers. This fibrillar network of collagen allows cartilage to entrap the proteoglycan aggregate as well as provide tensile strength to the tissue. Type II collagen, which adds structure and strength to connective tissues, is found primarily in cartilage, the jelly-like substance that fills the eyeball (the vitreous), the inner ear, and the center portion of the discs between the vertebrae in the spine (nucleus pulposus). Three pro-alpha1(II) chains twist together to form a triple-stranded, rope like procollagen molecule. These procollagen molecules must be processed by enzymes in the cell. Once these molecules are processed, they leave the cell and arrange themselves into long, thin fibrils that cross-link to one another in the spaces around cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.