Order Entry
Puerto Rico
ContactUsLinkComponent
Rabbit OC ELISA Kit
Rabbit OC ELISA Kit
Catalog # MSPP-SEA471RB
Supplier:  Cloud-Clone
CAS Number:  
undefined
Rabbit OC ELISA Kit
Catalog # MSPP-SEA471RB
Supplier:  Cloud-Clone
Supplier Number:  SEA471RB
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    OC
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Osteocalcin (OC) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.122 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA471RB
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rabbit OC (Osteocalcin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.122 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Osteocalcin (OC) is the most abundant noncollagenous protein in mature bone where it constitutes 1 to 2% of the total protein. Synthesized by osteoblasts, it is incorporated into the bone matrix. Osteocalcin is a 49-residue protein with three gamma-carboxyglutamic acid residues, at positions 17, 21 and 24. These three residues confer on it a very strong ability to bind to hydroxyapatite. Osteocalcin is a noncollagenous protein found in bone and dentin. It is secreted by osteoblasts and thought to play a role in mineralization and calcium ion homeostasis. It has been stipulated that osteocalcin may also function as a negative regulator of bone formation, although its exact role is unknown.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.