- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Rabbit
- Target protein:IL8
- Size:1 Kit
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Interleukin 8 (IL8) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:3.12 - 200 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:1.16 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA080RB
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Rabbit IL8 (Interleukin 8). The assay range is from 3.12 to 200 pg/ml (Sandwich kit) with a sensitivity of 1.16 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interleukin 8 (IL-8), a member of the neutrophil-specific CXC subfamily of chemokines, is a potent neutrophil chemotactic and activating factor. It is a primary inflammatory cytokine produced by many cells in response to proinflammatory stimuli such as IL-1, TNF, LPS and viruses. Its function is, in part, to attract neutrophils to the site of inflammation and to activate them. The IL-8 cDNA sequence predicts a protein of 99 amino acids. Removal of a 22-residue signal peptide generates a mature protein of 77 amino acids (~8 kDa). Further proteolysis of the N-terminal end leads to a variant form with 72 amino acids, full activation of IL-8 may require cleavage to the 72 amino acid form. IL-8 can form non-covalent dimers in solution, especially at high concentrations, but dimerization is not necessary for biological activity.