Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Rabbit IFNa ELISA Kit

96 wells, 1 Kit
Catalog # MSPP-SEA033RB
Marketsource
Supplier:   Cloud-Clone
 :  SEA033RB
Rabbit IFNa ELISA Kit
Catalog # MSPP-SEA033RB
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Rabbit IFNa ELISA Kit
Rabbit IFNa ELISA Kit

96 wells, 1 Kit

CAS Number:  
Price
$684.00
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Rabbit IFNa (Interferon Alpha). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Streuli et al. (1980) showed that at least 3 different IFN-alpha genes are expressed in man. Furthermore, study of genomic DNA revealed the presence of at least 8 IFN genes.,Nagata et al. (1980) found that the alpha-interferon genes are devoid of intervening sequences. Using radioactive probes from purified cDNA clones of interferons, Owerbach et al. (1981) located at least 8 leukocyte interferon genes and a fibroblast interferon gene on chromosome 9. Shows et al. (1982) found that the alpha- and beta-interferon genes are on 9p. The mapping to 9pter-q12 was accomplished by blot hybridization of cloned interferon cDNA to DNA from human-mouse cell hybrids with a translocation involving chromosome 9.

 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Specifications

Specifications

  • Size:
    1 Kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Rabbit
  • Detection Range:
    15.6 - 1000 pg/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEA033RB
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    5.7 pg/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    IFNa
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interferon Alpha (IFNa) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications