Order Entry
Puerto Rico
ContactUsLinkComponent
Rabbit CRP ELISA Kit
Rabbit CRP ELISA Kit
Catalog # MSPP-SEA821RB
CAS Number:  
undefined
Rabbit CRP ELISA Kit
Catalog # MSPP-SEA821RB
Supplier Number:  SEA821RB
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    CRP
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between C Reactive Protein (CRP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.78 - 50 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.28 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA821RB
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rabbit CRP (C Reactive Protein). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.28 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

C-reactive protein is a liver-derived pattern recognition molecule that is increased in inflammatory states. It rapidly increases within hours after tissue injury, and it is suggested that it is part of the innate immune system and contributes to host defense. Since cardiovascular disease is at least in part an inflammatory process, CRP has been investigated in the context of arteriosclerosis and subsequent vascular disorders. Based on multiple epidemiological and intervention studies, minor CRP elevation has been shown to be associated with future major cardiovascular risk.CRP is known for several decades as a nonspecific inflammation marker. High CRP levels are detected in human blood during bacterial, viral and other infections, as well as in noninfectious diseases such as rheumatic disorders and malignancies.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.