Order Entry
Puerto Rico
ContactUsLinkComponent
Pig uPA ELISA Kit
Pig uPA ELISA Kit
Catalog # MSPP-SEA140PO
Supplier:  Cloud-Clone
CAS Number:  
undefined
Pig uPA ELISA Kit
Catalog # MSPP-SEA140PO
Supplier:  Cloud-Clone
Supplier Number:  SEA140PO
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Pig
  • Target protein:
    uPA
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Plasminogen Activator, Urokinase (uPA) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.057 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA140PO
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Pig uPA (Plasminogen Activator Urokinase). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.057 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Urokinase-type plasminogen activator is a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. The protein encoded by this gene converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. This gene's proprotein is cleaved at a Lys-Ile bond by plasmin to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.