Order Entry
Puerto Rico
ContactUsLinkComponent
Pig TNFa ELISA Kit
Pig TNFa ELISA Kit
Catalog # MSPP-SEA133PO
CAS Number:  
undefined
Pig TNFa ELISA Kit
Catalog # MSPP-SEA133PO
Supplier Number:  SEA133PO
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Pig
  • Target protein:
    TNFa
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha (TNFa) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.1 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA133PO
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Pig TNFa (Tumor Necrosis Factor Alpha). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

TNFa  is synthesized as a 26 kDa, type II transmembrane protein that is 233 amino acids in length. It contains a 30 amino acid (aa) cytoplasmic domain, a 26 aa transmembrane segment, and a 177 aa extracellular region. TNFa is assembled intracellularly to form a transmembrane, non-covalently-linked homotrimeric protein. The 157 aa residue soluble form of TNFa (sTNF-αis released from the C-terminus of the transmembrane protein through the activity of TNFa-converting enzyme (TACE), a membrane -bound disintegrin metalloproteinase. Rat cells known to express TNF-αinclude B cells, colonic columnar epithelial cells, NK and CD3 CD56 hepatic natural T cells, macrophages, monocytes and monocyte-derived dendritic cells, CD4 and CD8 T cells, mast cells, neutrophils, keratinocytes, plasma cells, and adipocytes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.