Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Pig GM-CSF ELISA Kit

Pig GM-CSF ELISA Kit

96 wells, 1 Kit
Catalog # MSPP-SEA045PO
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEA045PO
Pig GM-CSF ELISA Kit
Catalog # MSPP-SEA045PO
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Pig GM-CSF ELISA Kit
Pig GM-CSF ELISA Kit

96 wells, 1 Kit

CAS Number:  
Price
$756.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Pig GM to CSF (Colony Stimulating Factor 2 Granulocyte Macrophage). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.0 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Granulocyte-macrophage colony stimulating factor (GM-CSF) was initially identified by its ability to stimulate the formation of granulocyte and macrophage colonies from hematopoietic progenitor cells in semi-solid media. in vivoand in vitro studies have now demonstrated that GM-CSF is a pleiotropic cytokine that induces myeloid precursor cells to proliferate and differentiate into neutrophils, monocyte/macrophages and eosinophils. GM-CSF has also been shown to stimulate the proliferation and differentiation of erythroid and megakaryocyte progenitor cells. In addition, GM-CSF is a survival factor for and activates the effector functions of mature neutrophils, monocytes/macrophages and eosinophils.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 Kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Pig
  • Detection Range:
    15.6 - 1000 pg/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEA045PO
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    6.0 pg/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    GM-CSF
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications